missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Cyanocyclohexylamine hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 15419388
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419388 1 g 1g
15408026 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419388 Supplier Thermo Scientific Alfa Aesar Supplier No. H62938.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 920966-30-9 | C7H13ClN2 | 160.645 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920966-30-9
Molecular Formula C7H13ClN2
Molecular Weight (g/mol) 160.645
MDL Number MFCD22548413
InChI Key ZBUXGIYHCTWUTA-ZJLYAJKPSA-N
Synonym trans-3-aminocyclohexanecarbonitrile hydrochloride, 1r,3r-3-aminocyclohexane-1-carbonitrile hydrochloride
PubChem CID 67278607
IUPAC Name (1R,3R)-3-aminocyclohexane-1-carbonitrile;hydrochloride
SMILES C1CC(CC(C1)N)C#N.Cl

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 160.64
Percent Purity 97%
Chemical Name or Material trans-3-Cyanocyclohexylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.