missing translation for 'onlineSavingsMsg'
Learn More

trans-3'-Hydroxy Cotinine N-Beta-D-Glucuronide, TRC

Product Code. 30008932
Change view
Click to view available options
Quantity:
1 mg
2 mg
5 mg
Unit Size:
1mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008932 1 mg 1mg
30132850 2 mg 2mg
30138139 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008932 Supplier Toronto Research Chemicals Supplier No. TRCH9245301MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146275-18-5
Molecular Formula C16 H20 N2 O8
Molecular Weight (g/mol) 368.34
Synonym Pyridinium, 1-β-D-glucopyranuronosyl-3-[(2S,4R)-4-hydroxy-1-methyl-5-oxo-2-pyrrolidinyl]-, inner salt (9CI, ACI), Pyridinium, 1-β-D-glucopyranuronosyl-3-(4-hydroxy-1-methyl-5-oxo-2-pyrrolidinyl)-, inner salt, (2S-trans)- (ZCI), trans-3'-Hydroxycotinine-N-glucuronide, trans-3'-Hydroxy Cotinine N-β-D-Glucuronide, 1-β-D-Glucopyranuronosyl-3-[(2S,4R)-4-hydroxy-1-methyl-5-oxo-2-pyrrolidinyl]pyridinium inner salt, (3R,5S)-3-Hydroxy-1-methyl-5-(3'-pyridinyl)-2-pyrrolidinone N'-β-D-Glucuronide, trans-3'-Hydroxycotinine N-β-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-[(3R,5S)-1-methyl-2-oxo-5-pyridin-3-ylpyrrolidin-3-yl]oxyoxane-2-carboxylic acid
SMILES CN1[C@@H](C[C@@H](O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)C1=O)c3cccnc3

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H20N2O8/c1-18-8(7-3-2-4-17-6-7)5-9(14(18)22)25-16-12(21)10(19)11(20)13(26-16)15(23)24/h2-4,6,8-13,16,19-21H,5H2,1H3,(H,23,24)/t8-,9+,10-,11-,12+,13-,16+/m0/s1
Formula Weight 368.122
Chemical Name or Material trans-3'-Hydroxycotinine-N-beta-D-glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.