missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Octene, 97%, Thermo Scientific Chemicals

Product Code. 11324838
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324838 2 g 2g
11304838 10 g 10g
11314838 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324838 Supplier Thermo Scientific Alfa Aesar Supplier No. 043154.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14919-01-8 | Molecular Formula: C8H16 | Molecular Weight: 112.216 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14919-01-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00039971
InChI Key YCTDZYMMFQCTEO-FNORWQNLSA-N
Synonym trans-3-octene, e-3-octene, e-oct-3-ene, 3-octene, e, 3-octene, 3e, unii-2a4p9ko860, 3-octene, 3e-3-octene, 3e-oct-3-ene, cis-3-octene
PubChem CID 638228
IUPAC Name (E)-oct-3-ene
SMILES CCCCC=CCC

Specifications

Density 0.716 g/mL
Boiling Point 121°C to 222°C
Flash Point 13°C (57°F)
Odor Characteristic
Refractive Index 1.413
Quantity 2 g
UN Number UN3295
Formula Weight 112.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material trans-3-Octene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.