missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde, 97%

Product Code. 15428046
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15428046

Brand: Thermo Scientific Alfa Aesar H62957.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 412357-50-7 | C15H19NO3 | 261.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 412357-50-7
Molecular Formula C15H19NO3
Molecular Weight (g/mol) 261.32
MDL Number MFCD03844593,MFCD06657671
InChI Key SOIRQISQWQDGSN-UHFFFAOYSA-N
PubChem CID 21882542
IUPAC Name benzyl N-(4-formylcyclohexyl)carbamate
SMILES O=CC1CCC(CC1)NC(=O)OCC1=CC=CC=C1

Specifications

Synonym benzyl 4-formylcyclohexylcarbamate, benzyl cis-4-formylcyclohexyl carbamate, benzyl cis-4-formylcyclohexylcarbamate, benzyl 4-formylcyclohexyl carbamate, trans-4-benzyloxycarbonylamino-cyclohexanecarbaldehyde, benzyl 1r,4r-4-formylcyclohexylcarbamate, trans-4-benzyloxycarbonylamino cyclohexanecarbaldehyde, benzyl n-4-formylcyclohexyl carbamate, benzyl4-formylcyclohexylcarbamate, soirqisqwqdgsn-mqmhxkeqsa-n
Quantity 1 g
Formula Weight 261.31
Percent Purity 97%
Chemical Name or Material trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.