missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Benzyloxycarbonylamino)cyclohexylamine, 97%

Product Code. 15437446
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437446 5 g 5g
15447446 250 mg 250mg
15427446 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15437446 Supplier Thermo Scientific Alfa Aesar Supplier No. H62444.06
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 149423-77-8 | C14H20N2O2 | 248.33 g/mol

trans-4-(Benzyloxycarbonylamino)cyclohexylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Benzyloxycarbonylamino)cyclohexylamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149423-77-8
Molecular Formula C14H20N2O2
Molecular Weight (g/mol) 248.33
MDL Number MFCD03839901,MFCD06657668,MFCD07772066
InChI Key JQVBZZUMWRXDSQ-UHFFFAOYSA-N
Synonym benzyl cis-4-aminocyclohexyl carbamate, benzyl trans-4-aminocyclohexyl carbamate, n-cbz-trans-1,4-cyclohexanediamine, cis-1-n-cbz-1,4-cyclohexyldiamine, cis-n-cbz-1,4-cyclohexanediamine, 4-amino-cyclohexyl-carbamic acid benzyl ester, benzyl cis-4-aminocyclohexylcarbamate, benzyl 4-aminocyclohexyl carbamate, benzyl 1r,4r-4-aminocyclohexyl carbamate, 1-n-cbz-trans-1,4-cyclohexyldiamine
PubChem CID 18650082
IUPAC Name benzyl N-(4-aminocyclohexyl)carbamate
SMILES NC1CCC(CC1)NC(=O)OCC1=CC=CC=C1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 248.32
Percent Purity 97%
Chemical Name or Material trans-4-(Benzyloxycarbonylamino)cyclohexylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.