missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Boc-amino)cyclohexanemethylamine, 97%

Product Code. 15468648
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15468648

Brand: Thermo Scientific Alfa Aesar H62062.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 177583-27-6 | C12H24N2O2 | 228.336 g/mol

trans-4-(Boc-amino)cyclohexanemethylamine is used as a raw material intermediates for pharmaceutical and chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexanemethylamine is used as a raw material intermediates for pharmaceutical and chemical.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177583-27-6
Molecular Formula C12H24N2O2
Molecular Weight (g/mol) 228.336
MDL Number MFCD01861810
InChI Key NVQFOBONHIXDOC-UHFFFAOYSA-N
Synonym tert-butyl trans-4-aminomethyl cyclohexyl carbamate, tert-butyl n-4-aminomethyl cyclohexyl carbamate, tert-butyl cis-4-aminomethylcyclohexylcarbamate, tert-butyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, tert-butyl trans-4-aminomethylcyclohexyl carbamate, tert-butyl cis-4-aminomethyl cyclohexyl carbamate, cis-4-boc-amino cyclohexylmethylamine, tert-butyl trans-4-aminomethylcyclohexylcarbamate, trans-4-boc-amino cyclohexylmethylamine, tert-butyl 4-aminomethyl cyclohexyl carbamate
PubChem CID 2756045
IUPAC Name tert-butyl N-[4-(aminomethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CN

Specifications

Solubility Information Slightly soluble in water.
Quantity 5 g
Formula Weight 228.33
Percent Purity 97%
Chemical Name or Material trans-4-(Boc-amino)cyclohexanemethylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.