missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Boc-aminomethyl)cyclohexylamine, 97%

Product Code. 15419078
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419078 1 g 1g
15439078 250 mg 250mg
15429078 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15419078 Supplier Thermo Scientific Alfa Aesar Supplier No. H62564.03
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 192323-07-2 | C12H24N2O2 | 228.34 g/mol

trans-4-(Boc-aminomethyl)cyclohexylamine a compound with potential for biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-aminomethyl)cyclohexylamine a compound with potential for biochemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192323-07-2
Molecular Formula C12H24N2O2
Molecular Weight (g/mol) 228.34
MDL Number MFCD06657670,MFCD12408630,MFCD22395596
InChI Key HMMYZMWDTDJTRR-UHFFFAOYSA-N
Synonym cis-4-boc-aminomethyl cyclohexylamine, tert-butyl trans-4-aminocyclohexylmethylcarbamate, tert-butyl trans-4-aminocyclohexyl methyl carbamate, tert-butyl cis-4-aminocyclohexyl methyl carbamate, trans-4-boc-aminomethyl cyclohexylamine, 4-boc-aminomethyl-cyclohexylamine, tert-butyl n-4-aminocyclohexyl methyl carbamate, tert-butyl 4-aminocyclohexyl methyl carbamate, tert-butyl trans-4-aminocyclohexyl methylcarbamate
PubChem CID 21882540
IUPAC Name tert-butyl N-[(4-aminocyclohexyl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1CCC(N)CC1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 228.33
Percent Purity 97%
Chemical Name or Material trans-4-(Boc-aminomethyl)cyclohexylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.