missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Cyanocyclohexylamine hydrochloride, 97%

Product Code. 15497516
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497516 250 mg 250mg
15459048 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497516 Supplier Thermo Scientific Alfa Aesar Supplier No. H62522.MD
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 873537-33-8 | C7H13ClN2 | 160.645 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in a cool and dry condition. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873537-33-8
Molecular Formula C7H13ClN2
Molecular Weight (g/mol) 160.645
MDL Number MFCD21643021
InChI Key BTLVSLNZSMIPCW-UHFFFAOYSA-N
Synonym trans-4-aminocyclohexanecarbonitrile hydrochloride, cis-4-aminocyclohexanecarbonitrile hydrochloride, 4-amino-cyclohexanecarbonitrile hydrochloride, 4-aminocyclohexanecarbonitrile hydrochloride, 4-aminocyclohexane-1-carbonitrile hydrochloride, c7h12n2.hcl, 1s,4s-4-aminocyclohexanecarbonitrile hydrochloride, 1r,4r-4-aminocyclohexane-1-carbonitrile hydrochloride, 1s,4s-4-aminocyclohexane-1-carbonitrile hydrochloride, cyclohexanecarbonitrile, 4-amino-, monohydrochloride, trans
PubChem CID 53407876
IUPAC Name 4-aminocyclohexane-1-carbonitrile;hydrochloride
SMILES C1CC(CCC1C#N)N.Cl

Specifications

Quantity 250 mg
UN Number UN3439
Formula Weight 160.64
Percent Purity 97%
Chemical Name or Material trans-4-Cyanocyclohexylamine hydrochloride
compliance-icons
Product Identifier
  • trans-4-Cyanocyclohexylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: trans-4-Cyanocyclohexylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.