missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxy-D-proline, 97%

Product Code. 11347249
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347249 1 g 1g
15423687 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347249 Supplier Thermo Scientific Alfa Aesar Supplier No. H27421.03
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3398-22-9 | C5H9NO3 | 131.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3398-22-9
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00065608
InChI Key PMMYEEVYMWASQN-UHFFFAOYNA-N
Synonym trans-4-hydroxy-d-proline, 2r,4s-4-hydroxypyrrolidine-2-carboxylic acid, h-d-hyp-oh, d-proline, 4-hydroxy-, 4s, 2r,4s-4-hydroxy-pyrrolidine-2-carboxylic acid, d-hydroxyproline, ambotzhaa1169, pubchem9966, d-hyp-oh
PubChem CID 440074
ChEBI CHEBI:27992
IUPAC Name (2R,4S)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES OC1CNC(C1)C(O)=O

Specifications

Melting Point ∽260°C (decomposition)
Quantity 1 g
Formula Weight 131.13
Percent Purity 97%
Chemical Name or Material trans-4-Hydroxy-D-proline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.