missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals trans-4-Hydroxy-L-proline, 99+%

Product Code. 10326470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
500g
Product Code. Quantity Packaging unitSize
10326470 10 g Glass bottle 10g
10607662 25 g Glass bottle 25g
10336470 100 g Glass bottle 100g
11926601 500 g Glass bottle 500g
4 options
This item is not returnable. View return policy

Product Code. 10326470

Brand: Thermo Scientific Chemicals 121780100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51-35-4 | C5H9NO3 | 131.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-35-4
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
InChI Key PMMYEEVYMWASQN-DMTCNVIQSA-N
Synonym trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro
PubChem CID 5810
ChEBI CHEBI:18095
IUPAC Name (2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES C1C(CNC1C(=O)O)O

Especificaciones

Melting Point 274.0°C to 275.0°C
Color White
Loss on Drying 0.2% (105°C, 3 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 10 g
Beilstein 22,191
Merck Index 15,4878
Specific Rotation Condition - 75.50 (20.00°C c=5,H2O)
Specific Rotation − 75.50
Solubility Information Solubility in water: 357.8g/L (20° c). Other solubilities: slightly soluble in alcohol, insoluble in ether
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Formula Weight 131.13
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material trans-4-Hydroxy-L-proline
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.