missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Isopropylcyclohexanecarboxylic Acid, TRC

Product Code. 30405926
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405926 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405926 Supplier Toronto Research Chemicals Supplier No. TRCI8244001G
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans-4-(1-methylethyl)cyclohexanecarboxylic acid
CAS 7077-05-6
Molecular Formula C10 H18 O2
Quantity 1 g
Synonym trans-4-(1-Methylethyl)cyclohexanecarboxylic Acid, Cyclohexanecarboxylic acid, 4-isopropyl-, trans- (8CI), trans-4-Isopropylcyclohexanecarboxylic acid, trans-p-Menthan-7-oic acid
InChI Formula InChI=1S/C10H18O2/c1-7(2)8-3-5-9(6-4-8)10(11)12/h7-9H,3-6H2,1-2H3,(H,11,12)/t8-,9-
SMILES CC(C)[C@@H]1CC[C@H](CC1)C(=O)O
IUPAC Name 4-propan-2-ylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol) 170.25
Formula Weight 170.1307
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.