missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Methylcyclohexylamine, 99%

Product Code. 15585906 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15585906 25 g 25g
15575906 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15585906 Supplier Thermo Scientific Acros Supplier No. 455920250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2523-55-9 | C7H15N | 113.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2523-55-9
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.2
InChI Key KSMVBYPXNKCPAJ-UHFFFAOYSA-N
Synonym 4-methylcyclohexylamine, trans-4-methylcyclohexylamine, 4-methylcyclohexanamine, trans-4-methylcyclohexanamine, cyclohexanamine, 4-methyl, cis-4-methylcyclohexylamine, p-methylcyclohexylamine, trans-4-methyl-cyclohexylamine, trans-4-methylcyclohexyl amine, 4-methyl-cyclohexylamine
PubChem CID 80604
IUPAC Name 4-methylcyclohexan-1-amine
SMILES CC1CCC(CC1)N

Specifications

Density 0.8500g/mL
Boiling Point 151.0°C to 154.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3C6H10NH2
Refractive Index 1.4500 to 1.5480
Quantity 25 g
Specific Gravity 0.85
Formula Weight 113.2
Percent Purity 99%
Chemical Name or Material trans-4-Methylcyclohexylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • trans-4-Methyl cylclohexyl amine, 99%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.