missing translation for 'onlineSavingsMsg'
Learn More

trans-5-(4-Chlorophenyl)-4-methyl-2-thiazolidinethione, TRC

Product Code. 30336044
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30336044 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336044 Supplier Toronto Research Chemicals Supplier No. TRCC172445100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans-5-(4-Chlorophenyl)-4-methyl-2-thiazolidinethione
CAS 86603-14-7
Molecular Formula C10H10ClNS2
Quantity 100 mg
Synonym (4R,5R)-5-(4-Chlorophenyl)-4-methylthiazolidine-2-thione
InChI Formula InChI=1S/C10H10ClNS2/c1-6-9(14-10(13)12-6)7-2-4-8(11)5-3-7/h2-6,9H,1H3,(H,12,13)/t6-,9+/m1/s1
SMILES C[C@H]1NC(=S)S[C@@H]1c2ccc(Cl)cc2
IUPAC Name (4R,5R)-5-(4-chlorophenyl)-4-methyl-1,3-thiazolidine-2-thione
Molecular Weight (g/mol) 243.77
Formula Weight 242.9943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.