missing translation for 'onlineSavingsMsg'
Learn More

trans-Bis(benzonitrile)dichloropalladium(II), Pd 27.1%, Thermo Scientific Chemicals

Product Code. 11386707
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386707 1 g 1g
11376707 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386707 Supplier Thermo Scientific Alfa Aesar Supplier No. 010006.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14220-64-5
Molecular Formula C14H10Cl2N2Pd
Molecular Weight (g/mol) 383.57
MDL Number MFCD00013123
InChI Key QQPSWGUJSPTOSH-UHFFFAOYSA-L
Synonym bis benzonitrile palladium chloride, bis benzonitrile palladium ii chloride, trans-bis benzonitrile palladium ii chloride, bis benzonitrile palladium ii chloride, 500mg, palladium 2+ bis benzonitrile dichloride
PubChem CID 10271322
IUPAC Name benzonitrile;palladium(2+);dichloride
SMILES Cl[Pd++](Cl)([N]#CC1=CC=CC=C1)[N]#CC1=CC=CC=C1

Specifications

Color White
Linear Formula PdCl2[(C6H5)(CN)]2
Quantity 1 g
UN Number UN3439
Solubility Information Soluble in acetone, chloroform
Formula Weight 383.57
Concentration or Composition (by Analyte or Components) Pd 27.1%
Physical Form Powder
Chemical Name or Material trans-Bis(benzonitrile)dichloropalladium(II)
compliance-icons
Product Identifier
  • trans-Bis(benzonitrile)dichloropalladium (II)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Bis(benzonitrile)palladium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.