missing translation for 'onlineSavingsMsg'
Learn More

trans-(+)-Chrysanthemic acid, 99+%

Product Code. 10175110 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10175110 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10175110 Supplier Thermo Scientific Acros Supplier No. 263635000
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4638-92-0 | C10H16O2 | 168.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4638-92-0
Melting Point 17.0°C to 21.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C10H16O2
Refractive Index 1.4768 to 1.4788
MDL Number MFCD01941572
Quantity 500 mg
Merck Index 15, 2255
Specific Rotation + 14.20
Synonym --trans-chrysanthemic acid, 1s,3s-2,2-dimethyl-3-2-methylprop-1-en-1-yl cyclopropanecarboxylic acid, chrysanthemic acid l-trans, l-trans-chrysanthemic acid, 1s-trans-2,2-dimethyl-3-2-methyl-1-propenyl cyclopropanecarboxylic acid, 1s-trans-2,2-dimethyl-3-2-methylprop-1-enyl cyclopropanecarboxylic acid, --trans-chrysanthemumsaeure, cyclopropanecarboxylic acid, 2,2-dimethyl-3-2-methylpropenyl-, 1s-trans, 4-09-00-00169 beilstein handbook reference, chrysanthemic acid dl-trans-form
Solubility Information Solubility in water: soluble.
InChI Key XLOPRKKSAJMMEW-JGVFFNPUSA-N
SMILES CC(=CC1C(C1(C)C)C(=O)O)C
IUPAC Name (1S,3S)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylic acid
Molecular Weight (g/mol) 168.23
PubChem CID 33607
ChEBI CHEBI:39102
Formula Weight 168.23
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material trans-(+)-Chrysanthemic acid, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • trans-(+)-Chrysanthemic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.