missing translation for 'onlineSavingsMsg'
Learn More

trans-Cinnamic Acid, 98+%

Product Code. 10052020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10052020 100 g Plastic bottle 100g
10123020 5 g Glass bottle 5g
10337670 500 g Plastic bottle 500g
10073613 2.5 kg Plastic drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10052020 Supplier Thermo Scientific Acros Supplier No. 158571000
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 140-10-3 | C9H8O2 | 148.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-10-3
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00004369
InChI Key WBYWAXJHAXSJNI-VOTSOKGWSA-N
Synonym cinnamic acid, trans-cinnamic acid, e-cinnamic acid, 3-phenylacrylic acid, trans-3-phenylacrylic acid, zimtsaeure, 2e-3-phenylprop-2-enoic acid, 3-phenylprop-2-enoic acid, 3-phenylpropenoic acid
PubChem CID 444539
ChEBI CHEBI:35697
IUPAC Name (E)-3-phenylprop-2-enoic acid
SMILES C1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 131.0°C to 136.0°C
Boiling Point 300.0°C
Color White
Assay Percent Range 98+%
Linear Formula C6H5CH=CHCO2H
Beilstein 09, 573
Merck Index 14, 2299
Solubility Information Solubility in water: 0.4g/L in water (20°C). Other solubilities: soluble in glacial acetic acid, 1g/6mL alcohol - 1g/5mL methanol, 1g/12mL chloroform, soluble in benzene, ether, acetone, oils, and carbon disulfide
Formula Weight 148.16
Percent Purity ≥98%
Density 1.2480g/mL
Quantity 100 g
Flash Point >100°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Specific Gravity 1.248
Chemical Name or Material trans-Cinnamic acid, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • trans-Cinnamic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.