missing translation for 'onlineSavingsMsg'
Learn More

trans-Cinnamoyl Chloride, TRC

Product Code. 30354736
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30354736 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354736 Supplier Toronto Research Chemicals Supplier No. TRCC596290500MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans-Cinnamoyl Chloride
CAS 102-92-1
Molecular Formula C9H7ClO
Quantity 500 mg
Synonym 3-Phenyl-2-propenoyl chloride, 3-Phenylacryloyl chloride, 3-Phenylpropenoyl chloride, Benzylideneacetyl chloride, Cinnamic acid chloride, Cinnamic chloride, NSC 4683, β-Phenylacryloyl chloride
InChI Formula InChI=1S/C9H7ClO/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H/b7-6+
SMILES ClC(=O)\C=C\c1ccccc1
IUPAC Name (E)-3-phenylprop-2-enoyl chloride
Molecular Weight (g/mol) 166.6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.