Learn More
trans-Dichlorobis(diethylsulfide)platinum(II), Pt 43.7%
Description
It is used in Hydrosilylation. Platinum(II) complexes of diethylsulfide are convenient and versatile precursor for the preparation of a variety of organoplatinum(II) derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 1 g |
| Solubility Information | Soluble in acetone,alcohol,ethyl acetate and benzene |
| Formula Weight | 446.37 |
| Concentration or Composition (by Analyte or Components) | Pt 43.7% |
| Physical Form | Crystalline |
| Chemical Name or Material | trans-Dichlorobis(diethylsulfide)platinum(II) |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.