missing translation for 'onlineSavingsMsg'
Learn More

trans-Indole-3-acrylic acid, 98+%

Product Code. 11430113
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11430113 1 g 1g
11440113 5 g 5g
11450113 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11430113

Brand: Thermo Scientific Alfa Aesar A12143.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29953-71-7 | C11H9NO2 | 187.198 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29953-71-7
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.198
MDL Number MFCD00005633
InChI Key PLVPPLCLBIEYEA-AATRIKPKSA-N
Synonym 3-indoleacrylic acid, trans-3-indoleacrylic acid, indole-3-acrylic acid, 3-indolylacrylic acid, indole-3beta-acrylic acid, 3-1h-indol-3-yl acrylic acid, indole-3-crylic acid, 2e-3-1h-indol-3-yl prop-2-enoic acid, trans-3-1h-indol-3-yl acrylic acid, indoleacrylic acid van
PubChem CID 5375048
IUPAC Name (E)-3-(1H-indol-3-yl)prop-2-enoic acid
SMILES C1=CC=C2C(=C1)C(=CN2)C=CC(=O)O

Specifications

Melting Point ∼185°C (decomposition)
Quantity 1 g
Beilstein 6317
Solubility Information Soluble in water.
Formula Weight 187.2
Percent Purity ≥98%
Chemical Name or Material trans-Indole-3-acrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.