Learn More
trans-Indole-3-acrylic acid, 98+%
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.
Solubility
Soluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | ∼185°C (decomposition) |
| Quantity | 5 g |
| Beilstein | 6317 |
| Solubility Information | Soluble in water. |
| Formula Weight | 187.2 |
| Percent Purity | ≥98% |
| Chemical Name or Material | trans-Indole-3-acrylic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.