missing translation for 'onlineSavingsMsg'
Learn More

trans-Phytoene (Technical grade), TRC

Product Code. 30132860
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132860 10 mg 10mg
30136707 5 mg 5mg
30011840 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132860 Supplier Toronto Research Chemicals Supplier No. TRCP39881010MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-04-5
Molecular Formula C40H64
Molecular Weight (g/mol) 544.94
Synonym 7,7',8,8',11,11',12,12'-Octahydro-ψ,ψ-carotene, 2,6,10,14,19,23,27,31-octamethyl-2,6,10,14,16,18,22,26,30-dotriacontanonaene, all-trans-7,7',8,8',11,11',12,12'-Octahydro-lycopene, (all-E)-Phytoene, all-trans-Phytoene
SMILES CC(C)[C@@H](C(=O)N[C@@H](Cc1ccccc1)C[C@H](O)[C@@H](N)Cc2ccccc2)N3C(=O)NCCC3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C40H64/c1-33(2)19-13-23-37(7)27-17-31-39(9)29-15-25-35(5)21-11-12-22-36(6)26-16-30-40(10)32-18-28-38(8)24-14-20-34(3)4/h11-12,19-22,27-30H,13-18,23-26,31-32H2,1-10H3/b12-11+,35-21+,36-22+,37-27+,38-28+,39-29+,40-30+
Formula Weight 544.5008
Chemical Name or Material trans-Phytoene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.