missing translation for 'onlineSavingsMsg'
Learn More

trans-Stilbene, 96%, Thermo Scientific Chemicals

Product Code. 10011920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10011920 500 g Plastic bottle 500g
10666172 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10011920 Supplier Thermo Scientific Acros Supplier No. 161045000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

trans-Stilbene, 96%, C14H12, CAS Number-103-30-0, bibenzylidine, trans-stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, alpha,beta-diphenylethylene, stilbene, trans-1,2-diphenylethene, bibenzal, e-stilbene, bibenzylidene, 500g, 95% min. (HPLC), 05,II,537

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-30-0
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
MDL Number MFCD00064300
InChI Key PJANXHGTPQOBST-VAWYXSNFSA-N
Synonym trans-stilbene, e-stilbene, stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, bibenzylidene, bibenzylidine, bibenzal, trans-1,2-diphenylethene, alpha,beta-diphenylethylene
PubChem CID 638088
ChEBI CHEBI:36007
IUPAC Name (E)-stilbene
SMILES C1=CC=C(C=C1)C=CC2=CC=CC=C2

Specifications

Melting Point 122°C to 126°C
Color White to Beige
Boiling Point 305°C to 307°C (744.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H5CH=CHC6H5
Quantity 500 g
Beilstein 05,II,537
Merck Index 15,8946
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in 90 parts cold alcohol,freely soluble in benzene and ether
Formula Weight 180.25
Percent Purity 96%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material trans-Stilbene, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.