missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-1,3-Butadiene-1,4-dicarboxylic acid, 98%

Product Code. 11396736
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11396736 5 g 5g
11386736 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396736 Supplier Thermo Scientific Alfa Aesar Supplier No. L03987.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3588-17-8 | C6H6O4 | 142.11 g/mol

trans,trans-1,3-Butadiene-1,4-dicarboxylic acid is a metabolite found in urine that determines biological exposure index for workers exposed to benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans,trans-1,3-Butadiene-1,4-dicarboxylic acid is a metabolite found in urine that determines biological exposure index for workers exposed to benzene.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3588-17-8
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00002702
InChI Key TXXHDPDFNKHHGW-ZPUQHVIOSA-N
Synonym trans,trans-muconic acid, muconic acid, e,e-muconic acid, 2,4-hexadienedioic acid, trans, trans-muconic acid, hexa-2,4-dienedioic acid, 2e,4e-hexa-2,4-dienedioic acid, 1,3-butadiene-1,4-dicarboxylic acid, 2,4-hexadienedioic acid, e,e, trans,trans-2,4-hexadienedioic acid
PubChem CID 5356793
ChEBI CHEBI:27036
IUPAC Name (2E,4E)-hexa-2,4-dienedioic acid
SMILES C(=CC(=O)O)C=CC(=O)O

Specifications

Melting Point ∽300°C (decomposition)
Quantity 5 g
Beilstein 1722248
Merck Index 14,6299
Solubility Information Soluble in dimethyl sulfoxide and methanol.
Formula Weight 142.11
Percent Purity ≥98%
Chemical Name or Material trans,trans-1,3-Butadiene-1,4-dicarboxylic acid
compliance-icons
Product Identifier
  • trans,trans-1,3-Butadiene-1,4-dicarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: trans,trans-1,3-Butadiene-1,4-dicarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.