missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-2,4-Decadienal, TRC

Product Code. 30137142
Change view
Click to view available options
Quantity:
5 mL
10 mL
25 mL
Unit Size:
10mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137142 10 mL 10mL
30137005 25 mL 25mL
17954825 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137142 Supplier Toronto Research Chemicals Supplier No. TRCD21050010ML
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25152-84-5
Molecular Formula C10 H16 O
Molecular Weight (g/mol) 152.23
Synonym 2,4-Decadienal, (2E,4E)-, 2,4-Decadienal, (E,E)- (8CI), (2E,4E)-2,4-Decadienal, (2E,4E)-Deca-2,4-dienal, (2E,4E)-Decadienal, (E,E)-2,4-Decadien-1-al, (E,E)-2,4-Decadienal, 2-trans,4-trans-Decadienal, trans,trans-2,4-Decadien-1-al, trans,trans-2,4-Decadienal, trans-2,trans-4-Decadienal
IUPAC Name (2E,4E)-deca-2,4-dienal
SMILES CCCCC\C=C\C=C\C=O

Specifications

Quantity 10 mL
InChI Formula InChI=1S/C10H16O/c1-2-3-4-5-6-7-8-9-10-11/h6-10H,2-5H2,1H3/b7-6+,9-8+
Formula Weight 152.1201
Chemical Name or Material trans,trans-2,4-Decadienal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.