missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-2,4-Hexadien-1-ol, 99%, stabilized

Product Code. 10701361 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10701361

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17102-64-6 | C6H10O | 98.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17102-64-6
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00002925
InChI Key MEIRRNXMZYDVDW-MQQKCMAXSA-N
Synonym sorbic alcohol, 2,4-hexadien-1-ol, sorbyl alcohol, trans,trans-2,4-hexadien-1-ol, 2,4-hexadienol, hexacose, hexakose, sorbinic alcohol, 1-hydroxy-2,4-hexadiene, 2e,4e-hexa-2,4-dien-1-ol
PubChem CID 641256
IUPAC Name (2E,4E)-hexa-2,4-dien-1-ol
SMILES C\C=C\C=C\CO

Specifications

Melting Point 30.5°C to 31.5°C
Color Colorless to Yellow
Density 0.8700g/mL
Boiling Point 80.0°C (12.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=CHCH=CHCH2OH
Refractive Index 1.4990 to 1.5010
Quantity 10 g
Merck Index 15,8848
Specific Gravity 0.87
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and oils
Stabilizer 0.1% alpha-tocopherol
Formula Weight 98.14
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material trans, trans-2, 4-Hexadien-1-ol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • trans,trans-2,4-Hexadien-1-ol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.