missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-Farnesol, 97%

Product Code. 15469009
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469009 25 g 25g
15449009 1 g 1g
15459009 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15469009 Supplier Thermo Scientific Alfa Aesar Supplier No. L14348.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 106-28-5 | C15H26O | 222.37 g/mol

trans,trans-Farnesol is an intermediate used in the biological synthesis of cholesterol from mevalonic acid. It is used in the perfume industry. Further, it is used in the preparation of terpenes such as squalenes, cembranoids and forskolin. It is also used in the synthesis of Cecropia juvenile hormone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans,trans-Farnesol is an intermediate used in the biological synthesis of cholesterol from mevalonic acid. It is used in the perfume industry. Further, it is used in the preparation of terpenes such as squalenes, cembranoids and forskolin. It is also used in the synthesis of Cecropia juvenile hormone.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-28-5
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.37
MDL Number MFCD00002918
InChI Key CRDAMVZIKSXKFV-YFVJMOTDSA-N
Synonym farnesol, trans,trans-farnesol, e,e-farnesol, trans-farnesol, 2e,6e-3,7,11-trimethyldodeca-2,6,10-trien-1-ol, 2-trans,6-trans-farnesol, 2e,6e-farnesol, all-trans-farnesol, polyprenol, farnesyl alcohol
PubChem CID 445070
ChEBI CHEBI:16619
IUPAC Name (2E,6E)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CO

Specifications

Density 0.886
Boiling Point 137°C (3 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.49
Quantity 25 g
Beilstein 1723039
Sensitivity Light sensitive
Merck Index 14,3937
Solubility Information Miscible with alcohol. Immiscible with water.
Formula Weight 222.37
Percent Purity 97%
Chemical Name or Material trans,trans-Farnesol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.