missing translation for 'onlineSavingsMsg'
Learn More

trans-trans-Muconic acid, MedChemExpress

Product Code. 16485566
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485566 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16485566 Supplier Medchem Express Supplier No. HY113247
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress trans-trans-Muconic acid is a urinary metabolite of benzene and has been used as a biomarker of exposure to benzene in human.

trans-trans-Muconic acid is a urinary metabolite of benzene and has been used as a biomarker of exposure to benzene in human.

Specifications

Chemical Name or Material trans-trans-Muconic acid
CAS 3588-17-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C6H6O4
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (351.84 mM; Need ultrasonic)
SMILES O=C(O)/C=C/C=C/C(O)=O
Molecular Weight (g/mol) 142.11
Formula Weight 142.11
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.