missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-Muconic Acid, TRC

Product Code. 30480686
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480686 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30480686 Supplier Toronto Research Chemicals Supplier No. TRCM79100150G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans,trans-Muconic Acid
CAS 3588-17-8
Purity Grade Notes HPLC
Molecular Formula C6 H6 O4
Quantity 50 g
Synonym 2,4-Hexadienedioic acid','(2E,4E)- (9CI','ACI), (2E,4E)-2,4-Hexadienedioic acid (ACI), 2,4-Hexadienedioic acid','(E,E)- (8CI), (E,E)-1,3-Butadiene-1,4-dicarboxylic acid, (E,E)-Muconic acid, NSC 66486, trans,trans-2,4-Hexadienedioic acid, trans,trans-Muconic acid
InChI Formula InChI=1S/C6H6O4/c7-5(8)3-1-2-4-6(9)10/h1-4H,(H,7,8)(H,9,10)/b3-1+,4-2+
SMILES OC(=O)\C=C\C=C\C(=O)O
IUPAC Name (2E,4E)-hexa-2,4-dienedioic acid
Molecular Weight (g/mol) 142.11
Formula Weight 142.0266
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.