missing translation for 'onlineSavingsMsg'
Learn More

trans,trans,trans-1,5,9-Cyclododecatriene, 97%, stab. with 30-50ppm 4-tert-butylcatechol

Product Code. 11397336
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11397336

Brand: Thermo Scientific Alfa Aesar L09317.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 676-22-2 | C12H18 | 162.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 676-22-2
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00003720
InChI Key ZOLLIQAKMYWTBR-MBKAWSJDSA-N
Synonym 1,5,9-cyclododecatriene, e,e,e, 1,5,9-cyclododecatriene
PubChem CID 20835579
IUPAC Name (5E,9E)-cyclododeca-1,5,9-triene
SMILES C1CC=CCCC=CCCC=C1

Specifications

Melting Point 32°C to 34°C
Density 0.88
Boiling Point 237°C to 238°C
Flash Point 73°C (163°F)
Odor Pungent
Quantity 25 g
UN Number UN1759
Beilstein 969880
Formula Weight 162.28
Percent Purity 97%
Chemical Name or Material trans,trans,trans-1,5,9-Cyclododecatriene, stabilized with 30 to 50ppm 4-tert-butylcatechol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.