missing translation for 'onlineSavingsMsg'
Learn More

trans-Zeatin, 97%

Product Code. 15494449
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
This item is not returnable. View return policy

Product Code. 15494449

Brand: Thermo Scientific Alfa Aesar J61972.MF

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Plant growth hormone that stimulates cell division | CAS: 1637-39-4 | C10H13N5O | 219.248 g/mol

trans-Zeatin regulates cell division, development, and nutrient processing. It acts as a high-affinity ligand for a number of receptors, including arabidopsis histidine kinases (AHKs), specifically AHK3, which is important in mediating plant development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Zeatin regulates cell division, development, and nutrient processing. It acts as a high-affinity ligand for a number of receptors, including arabidopsis histidine kinases (AHKs), specifically AHK3, which is important in mediating plant development.

Solubility
Soluble in DMSO and DMF

Notes
Store at -20°C. Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1637-39-4
Molecular Formula C10H13N5O
Molecular Weight (g/mol) 219.248
MDL Number MFCD00213654
InChI Key UZKQTCBAMSWPJD-FARCUNLSSA-N
Synonym trans-zeatin, zeatin, e-zeatin, zeatine, e-2-methyl-4-1h-purin-6-ylamino-2-buten-1-ol, trans-zeatin synthetic, unii-7i6ooj9gr6, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, 2e, 2e-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, e
PubChem CID 449093
ChEBI CHEBI:16522
IUPAC Name (E)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol
SMILES CC(=CCNC1=NC=NC2=C1NC=N2)CO

Specifications

Melting Point 208°C to 210°C
Color White
Quantity 50 mg
Beilstein 616241
Merck Index 14,10117
Solubility Information Soluble in DMSO and DMF
Formula Weight 219.25
Percent Purity 97%
Physical Form Powder
Chemical Name or Material trans-Zeatin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.