missing translation for 'onlineSavingsMsg'
Learn More

Transcrocetin, MedChemExpress

Product Code. 16439258
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16439258 10 mg 10mg
16401678 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16439258 Supplier Medchem Express Supplier No. HYN2072
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Transcrocetin (trans-Crocetin), extracted from saffron (Crocus sativus L.), acts as an NMDA receptor antagonist with high affinity. Transcrocetin (trans-Crocetin) is capable of crossing the blood-brain barrier and reach the central nervous system (CNS).

Transcrocetin (trans-Crocetin), extracted from saffron (Crocus sativus L.), acts as an NMDA receptor antagonist with high affinity. Transcrocetin (trans-Crocetin) is capable of crossing the blood-brain barrier and reach the central nervous system (CNS).

Specifications

Chemical Name or Material Transcrocetin
CAS 27876-94-4
Color Orange
Purity Grade Notes Research
Molecular Formula C20H24O4
Quantity 10 mg
Synonym trans-Crocetin
Solubility Information DMSO : 1 mg/mL (3.05 mM; Need ultrasonic and warming)
SMILES O=C(O)/C(C)=C/C=C/C(C)=C/C=C/C=C(C)/C=C/C=C(C)/C(O)=O
Molecular Weight (g/mol) 328.4
Formula Weight 328.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.