Learn More
Corning™ Transfer Cap with Dip Tube, Non-pyrogenic
Product Code. 31932512 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
3 L Erlenmeyer Flask
5 L Erlenmeyer Flask
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31932512 5 L Erlenmeyer Flask Pack of 4
31932497 3 L Erlenmeyer Flask Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31932512 Supplier Corning™ Supplier No. 11895
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
USP class VI compliant materials for reliable fluid handling.
  • Virgin high-density polypropylene, high density polyethylene
  • Heavy metal free (meets CONEG requirements)
  • Chemically resistant, heat sealable thermoplastic elastomer
  • Polycarbonate-silicone connector
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Class VI, CONEG Compliant
For Use With (Equipment) 5 L Erlenmeyer Flask
Product Type Transfer Cap