missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Transfer Vessels Dipper

Product Code. 10534064 Shop All Thermo Scientific Products
Change view
Click to view available options
:
12mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10534064 12mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10534064 Supplier Thermo Scientific™ Supplier No. AY509X6

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Remove small amounts of liquid nitrogen by using Thermo Scientific™ Transfer Vessels Dipper.

Remove small amounts of liquid nitrogen by using Thermo Scientific™ Transfer Vessels Dipper. This dipper is very convenient in the lab for liquid nitrogen storage sample preparation.

TRUSTED_SUSTAINABILITY

Specifications

Description LN2 dipper, 12mL
Type LN2 Vessel Dipper
Capacity (Metric) 12 mL
For Use With (Equipment) LN2 Transfer Vessels
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.