missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Multi-Channel Micropipette Transferpette™ -12 Electronic, DE-M
GREENER_CHOICE

Product Code. 10293212 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
1 to 20 μL
10 to 200 μL
15 to 300 μL
5 to 100 μL
Includes:
EU plug
Europe AC Adapter
UK/Ireland AC Adapter
UK/Ireland Adapter
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Includes Unit Size
10293212 15 to 300 μL Europe AC Adapter Each
12378623 15 to 300 μL UK/Ireland Adapter Each
12358623 5 to 100 μL UK/Ireland AC Adapter Each
11673229 10 to 200 μL UK/Ireland AC Adapter Each
10131932 5 to 100 μL Europe AC Adapter Each
11663229 1 to 20 μL EU plug Each
12634060 1 to 20 μL UK/Ireland Adapter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10293212 Supplier Brand™ Supplier No. 705456
  • ONLINE EXCLUSIVE PRICE
    1030.00 €
    985.40 € / Pack of 1
    Save 44.60 €
    4% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ergonomic, handy design. Clear and intuitive operation. Up to 4,000 pipetting cycles on a single charge.

  • Transferpette electronic pipettes combine the globally proven properties of mechanical piston operated air displacement pipettes by BRAND with the advantages of an electronic device.
  • Features: The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).
  • Functions: pipetting, reverse pipetting, mixing, dispensing, up to 300 μl also GEL-Electrophoresis.
    Easy Calibration-Technique for making quick adjustments of the Transferpette™ electronic, without tools.
  • Limitations of use: Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F) (other temperatures upon request)
  • Vapor pressure up to 500 mbar
  • Viscosity: 260 mPa s
  • For viscous media, the speed must be adjusted if necessary.

TRUSTED_SUSTAINABILITY

Specifications

Accuracy ≤ 0.6%, ± 1.8μL
Angle 360°
Autoclavable Autoclavable
Color Green
Number of Channels 12
Product Line Transferpette
Programmability Yes
Type Electronic Microliter Pipette
Volume (Metric) 15 to 300 μL
Display LCD
Finger Style Ergonomic
Includes Europe AC Adapter
Increments 0.5μL
Quantity 1 Each
Electrical Requirements 230V/50 Hz
Voltage 230 V
Frequency 50 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.