missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Multi-Channel Micropipette Transferpette™ -8 Electronic, DE-M

Product Code. 10477852 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 to 20 μL
10 to 200 μL
15 to 300 μL
5 to 100 μL
Color:
Gray
Green
Yellow
Includes:
Europe AC Adapter
UK/Ireland AC Adapter
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color Includes Unit Size
10477852 1 to 20 μL Gray Europe AC Adapter Each
12398603 1 to 20 μL Gray UK/Ireland AC Adapter Each
10797444 10 to 200 μL Yellow Europe AC Adapter Each
12328613 15 to 300 μL Green UK/Ireland AC Adapter Each
12308613 5 to 100 μL Yellow UK/Ireland AC Adapter Each
11613229 10 to 200 μL Yellow UK/Ireland AC Adapter Each
11603229 0.5 to 10 μL Gray UK/Ireland AC Adapter Each
10233172 15 to 300 μL Green Europe AC Adapter Each
10191972 5 to 100 μL Yellow Europe AC Adapter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10477852 Supplier BRAND™ Supplier No. 705400
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ergonomic, handy design. Clear and intuitive operation. Up to 4,000 pipetting cycles on a single charge.

Transferpette™ electronic pipettes combines the globally proven properties of mechanical piston operated air displacement pipettes by BRAND with the advantages of an electronic device.

  • Features:The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).
  • Functions: pipetting, reverse pipetting, mixing, dispensing, up to 300 μl also GEL-Electrophoresis.
  • Easy Calibration-Technique for making quick adjustments of the Transferpette™ electronic, without tools.
  • Limitations of use: Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F) (other temperatures upon request)
  • Vapor pressure up to 500 mbar
  • Viscosity: 260 mPa s
  • For viscous media, the speed must be adjusted if necessary.
TRUSTED_SUSTAINABILITY

Specifications

Accuracy ≤ 1.0%, ± 0.2μL
Angle 360°
Autoclavable Autoclavable
Color Gray
Model 8
Number of Channels 8
Product Line Transferpette
Programmability Yes
Type Electronic Microliter Pipette
Volume (Metric) 1 to 20 μL
Display LCD
Finger Style Ergonomic
Includes Europe AC Adapter
Increments 0.02μL
Quantity 1 Each
Electrical Requirements 230V/50 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.