missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brand™ Multi-Channel Micropipette Transferpette™ -8 electronic, 0.5-10 μL, DE-M
Shop All Brand ProductsDescription
Transferpette™ electronic pipettes combines the globally proven properties of mechanical piston operated air displacement pipettes by BRAND with the advantages of an electronic device.
- Features: The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).
- Functions: pipetting, reverse pipetting, mixing, dispensing, up to 300 μl also GEL-Electrophoresis.
- Easy Calibration-Technique for making quick adjustments of the Transferpette™ electronic, without tools.
- Limitations of use: Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F) (other temperatures upon request)
- Vapor pressure up to 500 mbar
- Viscosity: 260 mPa s
- For viscous media, the speed must be adjusted if necessary.
Specifications
Specifications
| Accuracy | ≤ 1.2%, ± 0.12μL |
| Angle | 360° |
| Autoclavable | Autoclavable |
| Color | Gray |
| Model | 8 |
| Number of Channels | 8 |
| Product Line | Transferpette |
| Programmability | Yes |
| Type | Electronic Microliter Pipette |
| Volume (Metric) | 0.5 to 10 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?