missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human CD4 siRNA
Blocks transcription and translation of CD4 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_000607 |
---|---|
Cell Lines Tested | Jurkat Lymphoblast T Cells. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | CD4 |
Gene ID (Entrez) | 920 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16099241
|
enQuireBio™
QX49-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089241
|
enQuireBio™
QX49-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000607 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
920 | |
CD4mut, T-cell surface glycoprotein CD4, T-cell co-receptor CD4 | |
Lyophilized |
Jurkat Lymphoblast T Cells. | |
CD4 | |
siRNA | |
Human |