missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human EGFR / ErbB1 / HER1 siRNA
Blocks transcription and translation of EGFR / ErbB1 / HER1 from Human.
Brand: enQuireBio™ QX18-2nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_005219 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
1956 | |
2 nmol | |
Human |
WT-EGFR-CHO EGFR overexpressing CHO cells and MDA-MB 468 Human Breast Cancer Cells. | |
EGFR | |
siRNA | |
ERBB1, HER1, ERBB, mENA, PIG61, NISBD2, Epidermal growth factor receptor, EGF-R | |
Lyophilized |