missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human PD-L1 / CD274 siRNA
Blocks transcription and translation of PD-L1 / CD274 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_054862 |
---|---|
Cell Lines Tested | Hep3b Human Hepatocellular Carinoma Cells, RKO Human Colon Cancer Cells, and Primary Dendritic Cells (DCs) isolated from PBMCs. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | CD274 |
Gene ID (Entrez) | 29126 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16099251
|
enQuireBio™
QX7-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089251
|
enQuireBio™
QX7-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_054862 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
29126 | |
PDL1, CD274, Programmed Cell Death 1 Ligand 1, PDCD1L1, PDCD1 Ligand 1, B7H1, B7-H1, PD-1 L1, PD-1 Ligand 1 | |
Lyophilized |
Hep3b Human Hepatocellular Carinoma Cells, RKO Human Colon Cancer Cells, and Primary Dendritic Cells (DCs) isolated from PBMCs. | |
CD274 | |
siRNA | |
Human |