missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Rat Nerve growth factor receptor / p75NTR siRNA
Blocks transcription and translation of Nerve growth factor receptor / p75NTR from Rat.
Brand: enQuireBio™ QX35-10nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_036742 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
24596 | |
10 nmol | |
Rat |
Pc12 Rat Pheocromocytoma Cells from the Adrenal Medulla and Rat Primary Neurons. | |
Ngfr | |
siRNA | |
p75NTR, Tnfrsf16, p75, RNNGFRR, LNGFR, low affinity nerve growth factor receptor, Tumor necrosis factor receptor superfamily member 16, NGFR | |
Lyophilized |