Learn More
Zeiss™ Transmitted Light Equipment S
Product Code. 10289589
Change view
Click to view available options
Type:
Transmitted Light Equipment
Unit Size:
Each
Product Code. Type Unit Size
10289589 Transmitted Light Equipment Each
1 options
Product Code. 10289589 Supplier Zeiss™ Supplier No. 4355010000000
  • ONLINE EXCLUSIVE PRICE
    2710.00 €
    2299.22 € / Pack of 1
    Save 410.78 €
    15% Off
Estimated Shipment: 11-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ergonomic console with light guide and two adjustable reflectors for bright field/dark field and for variable oblique light
Compatible Microscopes:
  • Axio Zoom.V16
  • SteREO Discovery.V12
  • SteREO Discovery.V20
  • SteREO Discovery.V8
  • SteREO Lumar.V12
Specifica
Type Transmitted Light Equipment
Description Transmitted light equipment
For Use With (Application) Transmitted Light
For Use With (Equipment) Stand Base 450
Includes Light guide and 2 shiftable and tiltable reflectors for bright field/dark field and for variable oblique light
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.