Learn More
Kartell™ HDPE General Purpose Funnels
Manufactured from HDPE for optimum strength and chemical resistance. Kartell™ HDPE General Purpose Funnels provide safety and precision in filling bottles in the laboratory.
Brand: Kartell™ 0017500
54.64 EUR valid until 2025-12-20
BEST PRICE on Promo! Use promo code "24125" to get your promotional price.
Description
Manufactured from HDPE for optimum strength and chemical resistance. Kartell™ HDPE General Purpose Funnels provide safety and precision in filling bottles in the laboratory.
- Suitable for a wide range of liquids
- Upstand on rim reduces potential overflow
- Outer ribbing eliminates back pressure
- Hanging eye for storage
Specifications
| 180mm | |
| 30mm | |
| Suitable for a wide range of liquids |
| 1500 mL | |
| HDPE | |
| 68.5mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.