Learn More
Kartell™ HDPE General Purpose Funnels
Manufactured from HDPE for optimum strength and chemical resistance. Kartell™ HDPE General Purpose Funnels provide safety and precision in filling bottles in the laboratory.
Brand:  Kartell™ 0017600
59.99 EUR valid until 2025-12-20
BEST PRICE on Promo! Use promo code "24125" to get your promotional price.
Product Code. 15723189
  • ONLINE EXCLUSIVE PRICE
    69.95 €
    68.25 € / Pack of 10
    Save 1.70 €
    2% Off
Only null left
Estimated Shipment: 27-01-2026
Add to basket
Description
Description
Manufactured from HDPE for optimum strength and chemical resistance. Kartell™ HDPE General Purpose Funnels provide safety and precision in filling bottles in the laboratory.
  • Suitable for a wide range of liquids
  • Upstand on rim reduces potential overflow
  • Outer ribbing eliminates back pressure 
  • Hanging eye for storage