missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Wash Bottles, Transparent Polypropylene

Product Code. 15752026
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Thread Size (Metric):
GL 25
GL 32
GL 45
GL 63
Unit Size:
Pack of 12
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Thread Size (Metric) Unit Size
15752026 1000 mL GL 32 Pack of 12
15712026 250 mL GL 45 Pack of 12
15742026 250 mL GL 25 Pack of 12
15722026 500 mL GL 45 Pack of 12
10746073 500 mL GL 25 Pack of 12
15732026 1000 mL GL 63 Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15752026 Supplier VITLAB™ Supplier No. 95193
  • ONLINE EXCLUSIVE PRICE
    115.00 €
    113.64 € / Pack of 12
    Save 1.36 €
    1% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Wash equipment and materials with a precise spray jet and optimized backflow. VITLAB™ Wash Bottles, Transparent Polypropylene feature a screw cap with an incorporated spray tube on a narrow- or wide-mouth bottle. Spray bottles are individually wrapped, autoclavable and recyclable.

Wash bottles are appropriate for cleaning, coating and dousing in any laboratory environment. Polypropylene bottles are ideal for a variety of applications, including use with a range of chemicals, solvents and temperatures. Spray bottles are individually wrapped in clearly labeled polyethylene bags.
  • Designed for precise spray control
    • Finely drawn tip creates a precise spray jet for fine control
    • Optimised medium backflow through the continuous spray tube
  • Choice of narrow- or wide-mouth bottles
    • Narrow-mouth, tapered bottles for easier handling
    • Wide-mouth, square-shouldered bottles for easy and safe filling
    • All bottles feature screw cap lids with GL threads for a strong, secure hold
  • Constructed of durable, transparent polypropylene
    • Chemically resistant for use with liquid or powdered reagents
    • Temperature resistant and autoclavable
    • Transparent material for easy identification and monitoring
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Color Transparent
Material Polypropylene
Quantity 12Pack
Thread Size (Metric) GL 32
Mouth Narrow
Height (Metric) 215 mm
Diameter (Metric) 92 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Product Type Wash Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.