Learn More
VITLAB™ Transparent PTFE Watch Glasses
Product Code. 10366091
Change view
Click to view available options
Diameter (Metric):
125 mm
Unit Size:
Each
Product Code. Diameter (Metric) Unit Size
10366091 125 mm Each
1 options
Product Code. 10366091 Supplier VITLAB™ Supplier No. 113497
  • ONLINE EXCLUSIVE PRICE
    48.55 €
    39.97 € / Pack of 1
    Save 8.58 €
    17% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Autoclavable Autoclavable
Material PTFE
Shape Round
Type Watch Glass
For Use With (Application) Used as Beaker Covers
Diameter (Metric) 125 mm