Learn More
Bochem™ Transport Basket
Product Code. 15126423
Change view
Click to view available options
Dimensions (L x W x H):
300 x 400 x 150mm
Unit Size:
Each
Product Code. Dimensions (L x W x H) Unit Size
15126423 300 x 400 x 150mm Each
1 options
Product Code. 15126423 Supplier Bochem™ Supplier No. 17302
  • ONLINE EXCLUSIVE PRICE
    538.00 €
    370.14 € / Pack of 1
    Save 167.86 €
    31% Off
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Bochem™ Transport Basket is a stackable metal mesh basket with handles.
  • 10 x 10 x 3mm mesh dimensions
  • Stainless Steel
Specifications
Certifications/Compliance EURO-STANDARD
Description Stackable
Material 18/10 E-Poli
Dimensions (L x W x H) 300 x 400 x 150mm