missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Transport Case for LB 9509

Product Code. 31908720 Shop All Berthold Technologies Products
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31908720 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908720 Supplier Berthold Technologies Supplier No. 32700
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A metal transport case providing space for the Junior and accessories is an ideal solution for outdoor use.

  • Rugged, impact‐resistant housing for maximum protection during transport
  • Shock‐absorbing foam interior to keep instruments securely in place
  • Reinforced corners and edges for extra durability
  • Secure dual‐latch locking system to prevent accidental opening
  • Lightweight construction without compromising stability

Specifications

Type Transport Case
Color Blue
For Use With (Equipment) Junior LB 9509 Portable Luminometer
Height (Metric) 230 mm
Width (Metric) 290 mm
Weight (Metric) 3.5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.