missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Transport Medium

Product Code. 16645562
Click to view available options
Volume (Metric):
100 mL
1 L
Unit Size:
600mL
Pack of 6
This item is not returnable. View return policy

Product Code. 16645562

Brand: Corning™ 25500CI

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Corning™ Transport Medium is an isotonic, neutral pH solution that contains protective proteins, antibiotics to prevent microbial growth and contamination

  • This isotonic, neutral pH solution contains protective proteins/antibiotic to prevent microbial growth, and anti-mycotic to prevent contamination by yeast and fungi
  • For research and further manufacturing purposes only
  • Not for direct use in humans or animals.
  • Formulated using the final ingredient concentrations outlined in the CDC's Preparation of Viral Transport Medium (SOP#: DSR-052-01)
  • Formulation contains Hanks' Balanced Salt Solution with Ca and Mg (without phenol red), 2% Heat Inactivated Fetal Bovine Serum, 100 μg/mL Gentamicin and 0.5 μg/mL Amphotericin B
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Formulated in accordance to the CDC's Preparation of Viral Transport Medium guidelines
For Use With (Application) Research, Manufacturing Applications
Quantity 6/Cs.
Storage Requirements 2°C to 8°C
Volume (Metric) 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.