missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Transport Medium
Corning™ Transport Medium is an isotonic, neutral pH solution that contains protective proteins, antibiotics to prevent microbial growth and contamination
145.15 € - 650.74 €
Specifications
Certifications/Compliance | Formulated in accordance to the CDC's Preparation of Viral Transport Medium guidelines |
---|---|
For Use With (Application) | Research, Manufacturing Applications |
Quantity | 6/Cs. |
Storage Requirements | 2°C to 8°C |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
16645562
|
Corning™
25-500-CI |
100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16681563
|
Corning™
25-500-CM |
1 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- This isotonic, neutral pH solution contains protective proteins/antibiotic to prevent microbial growth, and anti-mycotic to prevent contamination by yeast and fungi
- For research and further manufacturing purposes only
- Not for direct use in humans or animals.
- Formulated using the final ingredient concentrations outlined in the CDC's Preparation of Viral Transport Medium (SOP#: DSR-052-01)
- Formulation contains Hanks' Balanced Salt Solution with Ca and Mg (without phenol red), 2% Heat Inactivated Fetal Bovine Serum, 100 μg/mL Gentamicin and 0.5 μg/mL Amphotericin B
Specifications
Formulated in accordance to the CDC's Preparation of Viral Transport Medium guidelines | |
Research, Manufacturing Applications | |
6/Cs. | |
2°C to 8°C |