missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Radiation Hazard Trays

Available in general purpose, bio-hazard and radiation safety formats, spill trays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.01000kg

Product Code. 12337247

  • 135.00 € / Each
Estimated Shipment: 18-04-2024
to see stock.
Add to basket

Description

Description

  • Available in general purpose, biohazard and radiation safety formats
  • Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
  • All trays supplied with one free liner when purchased
  • Additional environmentally friendly APET liners are provided in packs of 25
Specifications

Specifications

Yellow
Plastic
54 cm
68 cm
Radiation Hazard Tray
Rectangular
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers